siRNA Details
| Gene Symbol: Hey2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013904.1 | NP_038932.1 | 1 | 155 | ||
| GenBank mRNA | AB093589.1 | - | 1 | 23 | ||
| AF172287.1 | - | 1 | 155 | |||
| AF173902.1 | - | 1 | 147 | |||
| AF232240.1 | - | 1 | 151 | |||
| AJ271867.1 | - | 1 | 155 | |||
| AK158000.1 | - | 1 | 156 | |||
| BC103575.1 | - | 1 | 95 | 40045790 | ||
| BC103576.1 | - | 1 | 98 | 40045791 | ||
| BC105651.1 | - | 1 | 95 | 40046419 | ||
| BC105652.1 | - | 1 | 98 | 40046420 | ||

Hey2
Genomic Map