siRNA Details
| Gene Symbol: Hey2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013904.1 | NP_038932.1 | 5 | 1107 | ||
| GenBank mRNA | AB093589.1 | - | 5 | 975 | ||
| AF172287.1 | - | 5 | 1107 | |||
| AF173902.1 | - | 5 | 1099 | |||
| AF232240.1 | - | 5 | 1103 | |||
| AJ271867.1 | - | 5 | 1107 | |||
| AK031506.1 | - | 5 | 954 | |||
| AK158000.1 | - | 5 | 1108 | |||
| BC103575.1 | - | 5 | 1047 | 40045790 | ||
| BC103576.1 | - | 5 | 1050 | 40045791 | ||
| BC105651.1 | - | 5 | 1047 | 40046419 | ||
| BC105652.1 | - | 5 | 1050 | 40046420 | ||

Hey2
Genomic Map