siRNA Details
Gene Symbol: Hgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159328.1 | NP_001152800.1 | 8, 9 | 743 | ||
NM_008244.3 | NP_032270.3 | 8, 9 | 743 | |||
GenBank mRNA | AK046299.1 | - | 8, 9 | 704 | ||
AK145031.1 | - | 8, 9 | 663 | |||
AK166441.1 | - | 8, 9 | 717 | |||
AK206792.1 | - | 1, 2 | 120 | |||
AK210546.1 | - | 1, 2 | 58 | |||
AK215004.1 | - | 1, 2 | 120 | |||
AK216932.1 | - | 1, 2 | 70 | |||
BC003239.1 | - | 8, 9 | 687 | 3497966 | ||
D50050.1 | - | 8, 9 | 693 |
