siRNA Details
| Gene Symbol: Hiat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008246.2 | NP_032272.2 | 11 | 1323 | ||
| GenBank mRNA | AK004779.1 | - | 11 | 1262 | ||
| AK041662.1 | - | 1 | 2610 | |||
| AK135865.1 | - | 11 | 1144 | |||
| AK158838.1 | - | 11 | 1324 | |||
| AK169626.1 | - | 11 | 1263 | |||
| AK188731.1 | - | 1 | 34 | |||
| BC118047.1 | - | 11 | 1143 | 40106779 | ||
| D88315.1 | - | 11 | 1196 | |||

Hiat1
Genomic Map