siRNA Details
Gene Symbol: Hipk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145824.1 | NP_001139296.1 | 12 | 3075 | ||
NM_010434.2 | NP_034564.2 | 12 | 3078 | |||
GenBank mRNA | AF077660.1 | - | 12 | 2832 | ||
AF170305.1 | - | 11 | 2542 | |||
AF305238.1 | - | 11 | 2540 | |||
AK140130.1 | - | 12 | 3181 | |||
BC131663.1 | - | 12 | 2710 | |||
BC131664.1 | - | 12 | 2706 | 40110751 | ||
X97572.1 | - | 2 | 296 |
