siRNA Details
| Gene Symbol: Hmgcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008256.4 | NP_032282.2 | 8 | 1432 | ||
| GenBank mRNA | AK004865.1 | - | 8 | 1430 | ||
| AK004902.1 | - | 8 | 1428 | |||
| AK098104.1 | - | 8 | 1428 | |||
| BC014714.1 | - | 8 | 1418 | 4216637 | ||
| BC024744.1 | - | 8 | 1416 | 5123821 | ||

Hmgcs2
Genomic Map