siRNA Details
| Gene Symbol: Hmmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013552.2 | NP_038580.2 | 7, 8 | 724 | ||
| GenBank mRNA | AF031932.1 | - | 7, 8 | 709 | ||
| AF079222.1 | - | 8, 9 | 673 | |||
| AK143123.1 | - | 7, 8 | 724 | |||
| AK163410.1 | - | 7, 8 | 667 | |||
| AK164410.1 | - | 7, 8 | 698 | |||
| BC021427.1 | - | 7, 8 | 722 | 5035341 | ||
| X64550.1 | - | 3, 4 | 337 | |||

Hmmr
Genomic Map