siRNA Details
| Gene Symbol: Hmmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013552.2 | NP_038580.2 | 11, 12 | 1336 | ||
| GenBank mRNA | AF031932.1 | - | 11, 12 | 1321 | ||
| AF079222.1 | - | 12, 13 | 1285 | |||
| AK143123.1 | - | 11, 12 | 1336 | |||
| AK163410.1 | - | 11, 12 | 1279 | |||
| BC021427.1 | - | 11, 12 | 1334 | 5035341 | ||
| X64550.1 | - | 7, 8 | 949 | |||

Hmmr
Genomic Map