siRNA Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008261.2 | NP_032287.2 | 6 | 821 | ||
| GenBank mRNA | AK143948.1 | - | 6 | 846 | ||
| BC039220.1 | - | 6 | 821 | 4238842 | ||
| D29015.1 | - | 6 | 748 | |||
| EF193391.1 | - | 6 | 620 | |||
| EF193393.1 | - | 6 | 646 | |||
| FJ608819.1 | - | 7 | 775 | |||
| FJ608820.1 | - | 7 | 775 | |||
| FJ608821.1 | - | 7 | 775 | |||

Hnf4a
Genomic Map