siRNA Details
| Gene Symbol: Hoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010449.4 | NP_034579.3 | 2 | 951 | ||
| GenBank mRNA | AK083575.1 | - | 2 | 952 | ||
| AK143943.1 | - | 3 | 743 | |||
| BC138097.1 | - | 2 | 885 | 8861116 | ||
| BC138098.1 | - | 2 | 888 | 8861117 | ||
| M22115.1 | - | 2 | 920 | |||

Hoxa1
Genomic Map