siRNA Details
Gene Symbol: Hoxd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010469.2 | NP_034599.2 | 6 | 1717 | ||
GenBank mRNA | AK135479.1 | - | 2 | 508 | ||
BC139206.1 | - | 2 | 609 | 8862228 | ||
BC139207.1 | - | 2 | 609 | 8862229 | ||
J03770.1 | - | 2 | 688 |
