siRNA Details
Gene Symbol: Hrh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010973.2 | NP_001010973.1 | 2 | 1073 | ||
NM_008286.2 | NP_032312.2 | 2 | 1073 | |||
GenBank mRNA | AB444633.1 | - | 1 | 570 | ||
AB444634.1 | - | 1 | 570 | |||
AF513034.1 | - | 1 | 570 | |||
AF513035.1 | - | 1 | 570 | |||
AK020259.1 | - | 2 | 1074 | |||
AK163760.1 | - | 2 | 1070 | |||
BC101958.1 | - | 1 | 568 | |||
BC103552.1 | - | 1 | 569 | |||
BC103553.1 | - | 1 | 569 | |||
BC103554.1 | - | 1 | 569 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
