siRNA Details
| Gene Symbol: Hspa1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013558.2 | NP_038586.2 | 2 | 1903 | ||
| GenBank mRNA | BC129894.1 | - | 2 | 1772 | 40111516 | |
| BC129895.1 | - | 2 | 1774 | 40111517 | ||
| D85732.1 | - | 2 | 1874 | |||
| EU622851.1 | - | 1 | 1730 | |||
| M32218.1 | - | 1 | 1730 | |||
| Gene Symbol: Gm10501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134269.1 | - | 2 | 380 | ||

Hspa1l
Genomic Map