siRNA Details
| Gene Symbol: Hsd11b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008289.2 | NP_032315.2 | 5 | 1162 | ||
| GenBank mRNA | AK155153.1 | - | 5 | 1164 | ||
| BC014753.1 | - | 5 | 1153 | 4235545 | ||
| BC066209.1 | - | 5 | 1109 | 6431285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd11b2| Gene Symbol: Hsd11b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008289.2 | NP_032315.2 | 5 | 1162 | ||
| GenBank mRNA | AK155153.1 | - | 5 | 1164 | ||
| BC014753.1 | - | 5 | 1153 | 4235545 | ||
| BC066209.1 | - | 5 | 1109 | 6431285 | ||