siRNA Details
| Gene Symbol: Hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010475.1 | NP_034605.1 | 5 | 685 | ||
| GenBank mRNA | AK019406.1 | - | 2 | 203 | ||
| AK143346.1 | - | 5 | 634 | |||
| BC125657.1 | - | 5 | 684 | 40130172 | ||
| BC125659.1 | - | 5 | 684 | 40130174 | ||
| X89627.1 | - | 5 | 685 | |||

Hsd17b1
Genomic Map