siRNA Details
| Gene Symbol: Htr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008313.4 | NP_032339.2 | 6 | 779 | ||
| GenBank mRNA | AJ011369.1 | - | 5 | 569 | ||
| AK141587.1 | - | 6 | 723 | |||
| AK141625.1 | - | 6 | 780 | |||
| BC132189.1 | - | 5 | 608 | |||
| BC132191.1 | - | 5 | 608 | |||
| BC145159.1 | - | 5 | 608 | |||
| BC145161.1 | - | 5 | 608 | |||
| Y09585.1 | - | 5 | 569 | |||
| Y09587.1 | - | 5 | 569 | |||
| Y09588.1 | - | 5 | 569 | |||
More Information
Set Membership: |
|

Htr4
Genomic Map