siRNA Details
| Gene Symbol: Slc6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010484.2 | NP_034614.2 | 13, 14 | 2003 | ||
| GenBank mRNA | AF013604.1 | - | 12, 13 | 1816 | ||
| AK008410.1 | - | 2, 3 | 203 | |||
| AY765218.1 | - | 2, 3 | 261 | |||
| BC108978.1 | - | 13, 14 | 1985 | 40047534 | ||
| BC108979.1 | - | 13, 14 | 1985 | 40047537 | ||
| X66119.1 | - | 12, 13 | 1597 | |||

Slc6a4
Genomic Map