siRNA Details
| Gene Symbol: Slc6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010484.2 | NP_034614.2 | 2 | 204 | ||
| GenBank mRNA | AF013604.1 | - | 1 | 17 | ||
| BC108978.1 | - | 2 | 186 | 40047534 | ||
| BC108979.1 | - | 2 | 186 | 40047537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a4| Gene Symbol: Slc6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010484.2 | NP_034614.2 | 2 | 204 | ||
| GenBank mRNA | AF013604.1 | - | 1 | 17 | ||
| BC108978.1 | - | 2 | 186 | 40047534 | ||
| BC108979.1 | - | 2 | 186 | 40047537 | ||