siRNA Details
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038698.1 | NP_001033787.1 | 5 | 801 | ||
NM_001163397.1 | NP_001156869.1 | 5 | 637 | |||
NM_001163399.1 | NP_001156871.1 | 5 | 887 | |||
NM_010488.4 | NP_034618.2 | 5 | 887 | |||
GenBank mRNA | AF041341.1 | - | 5 | 649 | ||
AK014133.1 | - | 5 | 655 | |||
AK079088.1 | - | 5 | 792 | |||
BC048159.1 | - | 5 | 860 | 5703005 | ||
BC051987.1 | - | 558 | ||||
BC052451.1 | - | 5 | 764 | 6409658 | ||
D31953.1 | - | 5 | 634 |
