siRNA Details
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038698.1 | NP_001033787.1 | 2 | 313 | ||
NM_001163397.1 | NP_001156869.1 | 2 | 149 | |||
NM_001163399.1 | NP_001156871.1 | 2 | 399 | |||
NM_010488.4 | NP_034618.2 | 2 | 399 | |||
GenBank mRNA | AF041341.1 | - | 2 | 161 | ||
AK014133.1 | - | 2 | 167 | |||
AK079088.1 | - | 2 | 304 | |||
BB648788.1 | - | 2 | 400 | |||
BC048159.1 | - | 2 | 372 | 5703005 | ||
BC051987.1 | - | 70 | ||||
BC052451.1 | - | 2 | 276 | 6409658 | ||
CF726962.1 | - | 2 | 149 | |||
CN663544.1 | - | 2 | 313 | |||
D31953.1 | - | 2 | 146 | |||
DQ437511.1 | - | 3 | 249 | |||
DQ460221.1 | - | 2 | 187 | |||
JX178292.1 | - | 2 | 86 |
