siRNA Details
| Gene Symbol: Hyal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008317.4 | NP_032343.2 | 2 | 919 | ||
| GenBank mRNA | AF011567.1 | - | 2 | 905 | ||
| AK028942.1 | - | 936 | ||||
| BC021636.1 | - | 2 | 920 | 3990455 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hyal1| Gene Symbol: Hyal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008317.4 | NP_032343.2 | 2 | 919 | ||
| GenBank mRNA | AF011567.1 | - | 2 | 905 | ||
| AK028942.1 | - | 936 | ||||
| BC021636.1 | - | 2 | 920 | 3990455 | ||