siRNA Details
| Gene Symbol: Ide | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031156.3 | NP_112419.3 | 2 | 384 | ||
| GenBank mRNA | AJ278422.1 | - | 2 | 404 | ||
| AK004972.2 | - | 2 | 384 | |||
| AK014703.1 | - | 304 | ||||
| AK128986.1 | - | 2 | 401 | |||
| BC041675.1 | - | 2 | 416 | 4951971 | ||

Ide
Genomic Map