siRNA Details
| Gene Symbol: Idua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008325.4 | NP_032351.2 | 5, 6 | 909 | ||
| NM_172997.1 | NP_766585.1 | 4, 5 | 768 | |||
| GenBank mRNA | AK031414.1 | - | 4, 5 | 563 | ||
| AK045330.1 | - | 3, 4 | 396 | |||
| AK155627.1 | - | 5, 6 | 910 | |||
| BC057935.1 | - | 4, 5 | 530 | 5327150 | ||
| L34111.1 | - | 4, 5 | 483 | |||

Idua
Genomic Map