siRNA Details
| Gene Symbol: Irgm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008326.1 | NP_032352.1 | 3 | 500 | ||
| GenBank mRNA | AK002545.1 | - | 3 | 522 | ||
| AK167558.1 | - | 2 | 354 | |||
| AK171743.1 | - | 3 | 481 | |||
| BC145957.1 | - | 3 | 312 | 40131260 | ||
| U19119.1 | - | 3 | 500 | |||

Irgm1
Genomic Map