siRNA Details
| Gene Symbol: Ifit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008332.3 | NP_032358.1 | 3 | 1051 | ||
| GenBank mRNA | AK038027.1 | - | 3 | 804 | ||
| AK042366.1 | - | 3 | 833 | |||
| AK150206.1 | - | 2 | 653 | |||
| AK150253.1 | - | 2 | 653 | |||
| BC050835.1 | - | 3 | 799 | 6513417 | ||
| CT010395.1 | - | 1 | 575 | |||
| U43085.1 | - | 2 | 626 | |||

Ifit2
Genomic Map