siRNA Details
| Gene Symbol: Igf1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010513.2 | NP_034643.2 | 11 | 2884 | ||
| GenBank mRNA | AF056187.1 | - | 11 | 2282 | ||
| AK137220.1 | - | 6 | 1013 | |||
| AK155884.1 | - | 10 | 1780 | |||
| BC138868.1 | - | 11 | 2314 | 8861890 | ||
| BC138869.1 | - | 11 | 2314 | 8861891 | ||

Igf1r
Genomic Map