siRNA Details
| Gene Symbol: Igf2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010515.2 | NP_034645.2 | 8 | 1154 | ||
| GenBank mRNA | BC150817.1 | - | 8 | 1065 | 9087728 | |
| BC171973.1 | - | 8 | 1065 | 9054259 | ||
| U04710.1 | - | 8 | 1110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igf2r| Gene Symbol: Igf2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010515.2 | NP_034645.2 | 8 | 1154 | ||
| GenBank mRNA | BC150817.1 | - | 8 | 1065 | 9087728 | |
| BC171973.1 | - | 8 | 1065 | 9054259 | ||
| U04710.1 | - | 8 | 1110 | |||