siRNA Details
| Gene Symbol: Il10rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008349.5 | NP_032375.2 | 4 | 522 | ||
| GenBank mRNA | AF440787.1 | - | 4 | 449 | ||
| AK040938.1 | - | 4 | 438 | |||
| AK134042.1 | - | 4 | 515 | |||
| AK160991.1 | - | 4 | 1477 | |||
| BC145791.1 | - | 4 | 522 | 40131094 | ||
| U53696.1 | - | 4 | 435 | |||

Il10rb
Genomic Map