siRNA Details
| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008353.2 | NP_032379.2 | 4 | 503 | ||
| GenBank mRNA | AK137577.1 | - | 4 | 504 | ||
| BC057212.1 | - | 4 | 486 | 5350447 | ||
| U23922.1 | - | 4 | 458 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il12rb1| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008353.2 | NP_032379.2 | 4 | 503 | ||
| GenBank mRNA | AK137577.1 | - | 4 | 504 | ||
| BC057212.1 | - | 4 | 486 | 5350447 | ||
| U23922.1 | - | 4 | 458 | |||