siRNA Details
Gene Symbol: Il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080147.1 | - | 10 | 1162 | ||
AK082889.1 | - | 10 | 1157 | |||
AK089984.1 | - | 10 | 1193 | |||
AK143292.1 | - | 10 | 1162 | |||
AK154675.1 | - | 10 | 1218 | |||
BC021472.1 | - | 6 | 616 | |||
BC052425.1 | - | 10 | 1185 | 6835500 | ||
BC059939.1 | - | 10 | 1153 | 6477224 | ||
S80963.1 | - | 10 | 1190 |
