siRNA Details
| Gene Symbol: Il15ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271497.1 | NP_001258426.1 | 3, 4 | 468 | ||
| NM_008358.2 | NP_032384.1 | 3, 4 | 531 | |||
| NM_133836.2 | NP_598597.1 | 3, 4 | 584 | |||
| GenBank mRNA | AY221616.1 | - | 3, 4 | 209 | ||
| AY221619.1 | - | 3 | 230 | |||
| BC022705.1 | - | 3, 4 | 341 | |||
| BC095982.1 | - | 1 | 259 | |||
| BC132231.1 | - | 2, 3 | 131 | |||
| BC132233.1 | - | 2, 3 | 131 | |||
| BC145348.1 | - | 2, 3 | 131 | |||

Il15ra
Genomic Map