siRNA Details
| Gene Symbol: Il1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010555.4 | NP_034685.1 | 4 | 581 | ||
| GenBank mRNA | AK132264.1 | - | 4 | 608 | ||
| BC032962.1 | - | 3 | 298 | 1282531 | ||
| CT010182.1 | - | 3 | 489 | |||
| X59769.1 | - | 4 | 581 | |||

Il1r2
Genomic Map