siRNA Details
| Gene Symbol: Il18r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008365.2 | NP_032391.1 | 11 | 1853 | ||
| GenBank mRNA | U43673.1 | - | 11 | 1853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il18r1| Gene Symbol: Il18r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008365.2 | NP_032391.1 | 11 | 1853 | ||
| GenBank mRNA | U43673.1 | - | 11 | 1853 | ||