siRNA Details
| Gene Symbol: Il2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008368.4 | NP_032394.1 | 10 | 1142 | ||
| GenBank mRNA | AK017288.1 | - | 10 | 1142 | ||
| AK035289.1 | - | 10 | 1095 | |||
| AK157572.1 | - | 10 | 1143 | |||
| AK169481.1 | - | 10 | 1123 | |||
| BC026869.1 | - | 10 | 1115 | 4015306 | ||
| BC099616.1 | - | 10 | 1116 | 30024496 | ||
| M28052.1 | - | 10 | 1099 | |||

Il2rb
Genomic Map