siRNA Details
| Gene Symbol: Il7r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008372.4 | NP_032398.3 | 2 | 282 | ||
| GenBank mRNA | AF078906.1 | - | 2 | 148 | ||
| AK040364.1 | - | 2 | 265 | |||
| AK040740.1 | - | 2 | 265 | |||
| AK041838.1 | - | 2 | 173 | |||
| AK042264.1 | - | 2 | 227 | |||
| AK154516.1 | - | 2 | 242 | |||
| BC089571.1 | - | 2 | 234 | 6758487 | ||
| BY776757.1 | - | 2 | 283 | |||
| M29697.1 | - | 2 | 195 | |||

Il7r
Genomic Map