siRNA Details
| Gene Symbol: Inpp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY007563.1 | - | 7 | 692 | ||
| BC028864.1 | - | 7 | 678 | 4457437 | ||
| CA315937.1 | - | 7 | 675 | |||
| CK390048.1 | - | 3 | 316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inpp5b| Gene Symbol: Inpp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY007563.1 | - | 7 | 692 | ||
| BC028864.1 | - | 7 | 678 | 4457437 | ||
| CA315937.1 | - | 7 | 675 | |||
| CK390048.1 | - | 3 | 316 | |||