siRNA Details
| Gene Symbol: Inpp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF040094.2 | - | 5 | 556 | ||
| AK004601.1 | - | 6 | 563 | |||
| AK004722.2 | - | 5 | 458 | |||
| AY007563.1 | - | 11 | 1288 | |||
| BC028864.1 | - | 11 | 1274 | 4457437 | ||
| BU610644.1 | - | 5 | 439 | |||
| BU921160.1 | - | 2 | 53 | |||
| CA320508.1 | - | 5 | 563 | |||

Inpp5b
Genomic Map