siRNA Details
| Gene Symbol: Inppl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122739.1 | NP_001116211.1 | 13 | 1638 | ||
| NM_010567.2 | NP_034697.2 | 13 | 1872 | |||
| GenBank mRNA | AF162781.1 | - | 13 | 1722 | ||
| AK133359.1 | - | 13 | 1599 | |||
| AK149873.1 | - | 13 | 1656 | |||
| BC063080.1 | - | 13 | 1568 | 6416946 | ||

Inppl1
Genomic Map