siRNA Details
| Gene Symbol: Invs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281977.1 | NP_001268906.1 | 10 | 1436 | ||
| NM_001281978.1 | NP_001268907.1 | 12 | 1714 | |||
| NM_010569.4 | NP_034699.3 | 10 | 1488 | |||
| GenBank mRNA | AF034860.3 | - | 10 | 1430 | ||
| AJ010902.1 | - | 10 | 1429 | |||
| AK041671.1 | - | 9 | 1255 | |||
| AK158766.1 | - | 12 | 1673 | |||
| BC126929.1 | - | 10 | 1325 | 40081431 | ||

Invs
Genomic Map