siRNA Details
| Gene Symbol: Irf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008391.4 | NP_032417.3 | 9 | 1063 | ||
| GenBank mRNA | AK087926.1 | - | 8 | 931 | ||
| AK132099.1 | - | 9 | 1076 | |||
| AK155025.1 | - | 9 | 1048 | |||
| BC006577.1 | - | 9 | 1056 | 3586586 | ||
| BC110666.1 | - | 9 | 1035 | 4918447 | ||
| J03168.1 | - | 9 | 1018 | |||

Irf2
Genomic Map