siRNA Details
| Gene Symbol: Irf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013674.1 | NP_038702.1 | 7 | 829 | ||
| GenBank mRNA | AK089319.1 | - | 7 | 848 | ||
| AK089819.1 | - | 7 | 851 | |||
| AK154138.1 | - | 7 | 853 | |||
| AK154315.1 | - | 7 | 841 | |||
| AK156562.1 | - | 7 | 851 | |||
| BC137713.1 | - | 7 | 817 | 8860732 | ||
| BC137714.1 | - | 7 | 817 | 8860733 | ||
| U11692.1 | - | 7 | 829 | |||
| U34307.1 | - | 7 | 823 | |||

Irf4
Genomic Map