siRNA Details
| Gene Symbol: Irs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010572.2 | NP_034702.2 | 1 | 2268 | ||
| GenBank mRNA | AK045172.1 | - | 1 | 16 | ||
| BC060235.1 | - | 1 | 2268 | |||
| BC150782.1 | - | 1 | 2342 | 9087693 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irs4| Gene Symbol: Irs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010572.2 | NP_034702.2 | 1 | 2268 | ||
| GenBank mRNA | AK045172.1 | - | 1 | 16 | ||
| BC060235.1 | - | 1 | 2268 | |||
| BC150782.1 | - | 1 | 2342 | 9087693 | ||