siRNA Details
Gene Symbol: Irx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010573.2 | NP_034703.2 | 2 | 1396 | ||
GenBank mRNA | AF165984.1 | - | 2 | 1161 | ||
AK133937.1 | - | 2 | 1396 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Irx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010573.2 | NP_034703.2 | 2 | 1396 | ||
GenBank mRNA | AF165984.1 | - | 2 | 1161 | ||
AK133937.1 | - | 2 | 1396 |