siRNA Details
| Gene Symbol: Itga2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010575.2 | NP_034705.2 | 18, 19 | 2051 | ||
| GenBank mRNA | AF170316.1 | - | 18, 19 | 1912 | ||
| AK048700.1 | - | 13, 14 | 1559 | |||
| AK140749.1 | - | 13, 14 | 1703 | |||
| AK142289.1 | - | 18, 19 | 1913 | |||
| AK154619.1 | - | 18, 19 | 2052 | |||
| AK156872.1 | - | 17, 18 | 2233 | |||
| AK157012.1 | - | 17, 18 | 2236 | |||
| AK158443.1 | - | 18, 19 | 1903 | |||
| BC120493.1 | - | 18, 19 | 1901 | 40129416 | ||
| BC137570.1 | - | 18, 19 | 1901 | 8860589 | ||

Itga2b
Genomic Map