siRNA Details
| Gene Symbol: Itga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC053031.1 | - | 7 | 1518 | 6401146 | |
| BC062205.1 | - | 6 | 1293 | 4950774 | ||
| BC096741.1 | - | 8 | 1496 | |||
| D13867.1 | - | 7 | 1140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itga3| Gene Symbol: Itga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC053031.1 | - | 7 | 1518 | 6401146 | |
| BC062205.1 | - | 6 | 1293 | 4950774 | ||
| BC096741.1 | - | 8 | 1496 | |||
| D13867.1 | - | 7 | 1140 | |||