siRNA Details
| Gene Symbol: Itga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010576.3 | NP_034706.3 | 1 | 295 | ||
| GenBank mRNA | AB850880.1 | - | 1 | 60 | ||
| AF109136.1 | - | 1 | 131 | |||
| AK039959.1 | - | 1 | 149 | |||
| AK041096.1 | - | 1 | 295 | |||
| AK051676.1 | - | 1 | 221 | |||
| AK156015.1 | - | 1 | 191 | |||
| AK156517.1 | - | 1 | 65 | |||
| BC068313.1 | - | 1 | 75 | 30093248 | ||
| X53176.1 | - | 1 | 218 | |||

Itga4
Genomic Map