siRNA Details
| Gene Symbol: Itga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010576.3 | NP_034706.3 | 23 | 2683 | ||
| GenBank mRNA | AB850880.1 | - | 23 | 2448 | ||
| AF109136.1 | - | 23 | 2519 | |||
| BC068313.1 | - | 23 | 2463 | 30093248 | ||
| X53176.1 | - | 23 | 2606 | |||

Itga4
Genomic Map