siRNA Details
| Gene Symbol: Itga6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277970.1 | NP_001264899.1 | 14 | 2109 | ||
| NM_008397.4 | NP_032423.2 | 14 | 2109 | |||
| GenBank mRNA | AK034186.1 | - | 14 | 1966 | ||
| AK045391.1 | - | 7 | 690 | |||
| AK159653.1 | - | 9 | 1045 | |||
| BC049132.1 | - | 15 | 2034 | |||
| BC058095.1 | - | 15 | 2034 | 6826609 | ||
| X69902.1 | - | 14 | 2090 | |||

Itga6
Genomic Map