siRNA Details
| Gene Symbol: Itgae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008399.2 | NP_032425.2 | 24 | 2877 | ||
| NM_172944.2 | NP_766532.1 | 24 | 2877 | |||
| GenBank mRNA | AK040983.1 | - | 24 | 2878 | ||
| AK154021.1 | - | 24 | 2875 | |||
| BC150690.1 | - | 24 | 2877 | 9087601 | ||
| DQ386608.1 | - | 24 | 2877 | |||
| U12236.1 | - | 24 | 2901 | |||

Itgae
Genomic Map