siRNA Details
| Gene Symbol: Itgam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082960.1 | NP_001076429.1 | 6 | 579 | ||
| NM_008401.2 | NP_032427.2 | 6 | 579 | |||
| GenBank mRNA | AK154224.1 | - | 6 | 499 | ||
| AK154474.1 | - | 6 | 533 | |||
| AK154798.1 | - | 6 | 495 | |||
| AK155006.1 | - | 6 | 509 | |||
| AK155520.1 | - | 6 | 504 | |||
| AK155671.1 | - | 6 | 545 | |||
| AK155717.1 | - | 6 | 537 | |||
| AK170324.1 | - | 6 | 551 | |||
| AK171068.1 | - | 3 | 208 | |||
| AK171150.1 | - | 6 | 514 | |||

Itgam
Genomic Map